Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005973.4 | NP_005964.3 | 1 | 720 | ||
XM_005245313.2 | XP_005245370.1 | 1 | 720 | |||
XM_005245314.1 | XP_005245371.1 | 1 | 720 | |||
XM_011509726.1 | XP_011508028.1 | 1 | 720 | |||
GenBank mRNA | AK289872.1 | - | 1 | 660 | ||
BC004913.2 | - | 1 | 636 | 3535413 | ||
BC010450.1 | - | 1 | 648 | 4179105 | ||
BM423590.1 | - | 706 | ||||
KU178273.1 | - | 1 | 430 | |||
KU178274.1 | - | 1 | 430 | |||
KU178275.1 | - | 1 | 430 | |||
X97124.1 | - | 1 | 620 |