Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199283.1 | NP_001186212.1 | 6, 7 | 938 | ||
NM_001199284.1 | NP_001186213.1 | 6, 7 | 720 | |||
NM_173388.2 | NP_775564.1 | 6, 7 | 683 | |||
GenBank mRNA | AB120363.1 | - | 6, 7 | 716 | ||
AK143773.1 | - | 6, 7 | 781 | |||
AK159144.1 | - | 6, 7 | 906 | |||
BC042513.1 | - | 6, 7 | 679 | 3964020 |