Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173391.3 | NP_775567.2 | 11 | 1716 | ||
GenBank mRNA | AY090565.1 | - | 11 | 1716 |