Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173400.2 | NP_775576.2 | 5 | 720 | ||
GenBank mRNA | AK040101.1 | - | 5 | 723 | ||
AK051016.1 | - | 5 | 741 | |||
AK077681.1 | - | 5 | 740 | |||
AK169222.1 | - | 5 | 722 | |||
AK173206.1 | - | 5 | 692 | |||
BC068237.1 | - | 5 | 720 | 30550558 |