Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173437.2 | NP_775613.2 | 6 | 1859 | ||
GenBank mRNA | AF307453.1 | - | 6 | 2219 | ||
AK053255.1 | - | 4 | 838 | |||
AK082667.1 | - | 4 | 772 | |||
AK122458.1 | - | 8 | 2097 | |||
AK140435.1 | - | 6 | 1834 | |||
BC059840.1 | - | 6 | 2005 | 6836774 |