Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110516.1 | NP_001103986.1 | 10 | 1527 | ||
GenBank mRNA | AK149268.1 | - | 10 | 1527 | ||
AK161035.1 | - | 9 | 1541 | |||
AY458844.1 | - | 9 | 1344 | |||
BC026511.1 | - | 5 | 1415 | |||
BC026991.1 | - | 9 | 1649 | 5363470 |