Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl14ep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025102.1 | NP_001020273.1 | 5 | 1341 | ||
NM_173750.2 | NP_776111.1 | 5 | 1295 | |||
GenBank mRNA | AA407700.1 | - | 1 | 282 | ||
AK076781.1 | - | 5 | 1296 | |||
AK134965.1 | - | 5 | 1332 | |||
BC054106.1 | - | 5 | 1327 | 30021622 |