Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgalsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173752.4 | NP_776113.1 | 5 | 507 | ||
GenBank mRNA | AK041053.1 | - | 5 | 422 | ||
AK081513.1 | - | 2 | 192 | |||
AK156350.1 | - | 5 | 635 | |||
AK167557.1 | - | 5 | 445 | |||
BC019131.1 | - | 5 | 478 | 5036749 |