Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A730017C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167925.2 | NP_001161397.2 | 3 | 265 | ||
NM_173759.5 | NP_776120.3 | 2 | 214 | |||
GenBank mRNA | AK048493.1 | - | 3 | 1243 | ||
AK080452.1 | - | 4 | 218 | |||
AK086069.1 | - | 1 | 1326 | |||
BC075669.1 | - | 4 | 174 | 30550997 | ||
BY124558.1 | - | 4 | 285 | |||
BY125990.1 | - | 2 | 69 |