Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293560.1 | NP_001280489.1 | 12 | 1313 | ||
NM_173763.4 | NP_776124.1 | 12 | 1317 | |||
GenBank mRNA | AF363737.1 | - | 12 | 1287 | ||
AK049569.1 | - | 12 | 1286 | |||
AK145623.1 | - | 12 | 1311 | |||
BC037135.1 | - | 8 | 894 | |||
BC085287.1 | - | 13 | 1410 | |||
BC131942.1 | - | 11 | 1102 | 40130219 | ||
BC132615.1 | - | 11 | 1102 | 40130892 |