Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006004.3 | NP_005995.2 | 2, 3 | 231 | ||
GenBank mRNA | AK311967.1 | - | 2, 3 | 127 | ||
BC001426.2 | - | 2, 3 | 131 | 3140323 | ||
BC001934.1 | - | 2, 3 | 136 | 2823237 | ||
BC015177.1 | - | 2, 3 | 134 | 3905169 | ||
BC107703.1 | - | 2, 3 | 137 | 6605146 | ||
BT007070.1 | - | 2, 3 | 77 | |||
M36647.1 | - | 2, 3 | 107 | |||
Y00764.1 | - | 2, 3 | 113 |
Gene Symbol: UQCRHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001089591.1 | NP_001083060.1 | 1 | 127 |