Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175448.3 | NP_780657.1 | 3 | 1109 | ||
GenBank mRNA | AK039308.1 | - | 3 | 1110 | ||
AK049896.1 | - | 3 | 670 | |||
AK049988.1 | - | 3 | 678 | |||
AK053797.1 | - | 3 | 1132 | |||
AK139409.1 | - | 3 | 1184 | |||
AK139868.1 | - | 3 | 1162 | |||
AK163214.1 | - | 3 | 1130 | |||
AK169009.1 | - | 3 | 1279 | |||
BC058539.1 | - | 3 | 1116 | 5690124 |