Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175448.3 | NP_780657.1 | 6 | 1961 | ||
GenBank mRNA | AK039308.1 | - | 6 | 1962 | ||
AK049896.1 | - | 6 | 1522 | |||
AK139409.1 | - | 6 | 2036 | |||
AK139868.1 | - | 6 | 2015 | |||
AK169009.1 | - | 6 | 2131 |