Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175454.2 | NP_780663.1 | 16 | 2117 | ||
GenBank mRNA | AB087506.1 | - | 16 | 2117 | ||
AK044395.1 | - | 16 | 2095 | |||
AK049861.1 | - | 16 | 2116 | |||
AK050531.1 | - | 3 | 386 | |||
AK138882.1 | - | 16 | 2121 | |||
BC024617.1 | - | 16 | 2117 | 4190480 |