Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175455.4 | NP_780664.2 | 5 | 1043 | ||
GenBank mRNA | AK078203.1 | - | 5 | 1117 | ||
AK090387.1 | - | 5 | 1046 | |||
AK138465.1 | - | 4 | 1229 | |||
BC096609.1 | - | 5 | 919 | |||
BC119312.1 | - | 5 | 905 | 8734061 | ||
BC119314.1 | - | 5 | 905 | 8734063 |