Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270399.1 | NP_001257328.1 | 4 | 2255 | ||
NM_001270400.1 | NP_001257329.1 | 4 | 1783 | |||
NM_006009.3 | NP_006000.2 | 4 | 1695 | |||
GenBank mRNA | AF141347.1 | - | 4 | 1429 | ||
AF141348.1 | - | 1 | 982 | |||
AK057842.1 | - | 4 | 1449 | |||
AK289483.1 | - | 4 | 1622 | |||
AK297775.1 | - | 3 | 642 | |||
AK298838.1 | - | 4 | 1305 | |||
BC006468.1 | - | 4 | 1404 | 3528520 | ||
BC050637.2 | - | 4 | 1434 | 6050536 | ||
K00557.1 | - | 2 | 1023 |