Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam78b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160261.1 | NP_001153733.1 | 2 | 1325 | ||
NM_001160262.1 | NP_001153734.1 | 2 | 1325 | |||
NM_175461.4 | NP_780670.2 | 2 | 1325 | |||
GenBank mRNA | AK033840.1 | - | 1 | 823 | ||
AK047703.1 | - | 2 | 1163 | |||
AK049031.1 | - | 1 | 597 | |||
BC066094.1 | - | 2 | 1153 | |||
BC132161.1 | - | 3 | 1248 | 40130438 | ||
BC132525.1 | - | 2 | 1310 | 40130802 |