Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5rl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175471.2 | NP_780680.1 | 2 | 465 | ||
GenBank mRNA | AK053031.1 | - | 5 | 869 | ||
AK220154.1 | - | 3 | 367 | |||
AY437136.1 | - | 295 | ||||
BC004750.1 | - | 2 | 304 | |||
BC043687.1 | - | 2 | 465 | 5136046 |