Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177713.1 | NP_001171184.1 | 6 | 1230 | ||
NM_175475.3 | NP_780684.1 | 6 | 1281 | |||
GenBank mRNA | AK028272.1 | - | 4 | 765 | ||
AK137124.1 | - | 5 | 1556 | |||
AK166211.1 | - | 5 | 819 | |||
AY134662.1 | - | 6 | 1217 | |||
BC059246.1 | - | 6 | 1230 | 6400154 |