Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006015.4 | NP_006006.3 | 3 | 1776 | ||
NM_139135.2 | NP_624361.1 | 3 | 1776 | |||
GenBank mRNA | AB001895.1 | - | 2 | 541 | ||
AB210022.1 | - | 3 | 365 | |||
AF219114.1 | - | 3 | 365 | |||
AF231056.1 | - | 3 | 1781 | |||
AF265208.1 | - | 3 | 331 | |||
AF268913.1 | - | 2 | 254 | |||
AF521670.1 | - | 3 | 545 |