Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142401.1 | NP_001135873.1 | 2 | 376 | ||
NM_001142402.1 | NP_001135874.1 | 2 | 376 | |||
NM_001142403.1 | NP_001135875.1 | 2 | 376 | |||
NM_001142404.1 | NP_001135876.1 | 2 | 376 | |||
NM_006016.4 | NP_006007.2 | 2 | 376 | |||
GenBank mRNA | AF106518.1 | - | 2 | 195 | ||
AF263279.1 | - | 2 | 195 | |||
AF299341.1 | - | 2 | 195 | |||
AF299342.1 | - | 2 | 273 | |||
AF299343.1 | - | 2 | 273 | |||
AK298684.1 | - | 2 | 228 | |||
AK301692.1 | - | 2 | 286 | |||
AK303525.1 | - | 2 | 228 | |||
AK307193.1 | - | 2 | 284 | |||
AK308664.1 | - | 1 | 350 | |||
AK310854.1 | - | 2 | 217 | |||
AK312357.1 | - | 2 | 284 | |||
AK315908.1 | - | 2 | 227 | |||
BC011522.2 | - | 2 | 276 | 3854912 | ||
BC037914.1 | - | 1 | 878 | |||
D14043.1 | - | 2 | 274 | |||
DC403839.1 | - | 2 | 376 | |||
FJ200494.1 | - | 2 | 195 |