Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177337.3 | NP_796311.2 | 2 | 955 | ||
GenBank mRNA | AK008804.3 | - | 2 | 864 | ||
AK050053.1 | - | 2 | 866 | |||
AK155765.1 | - | 1 | 1577 | |||
BC064093.1 | - | 2 | 955 | 864589 |