Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177347.2 | NP_796321.1 | 1 | 750 | ||
GenBank mRNA | AY190047.1 | - | 1 | 750 |