Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164785.1 | NP_001158257.1 | 6 | 1215 | ||
NM_001164786.1 | NP_001158258.1 | 6 | 1215 | |||
NM_177431.4 | NP_803180.3 | 6 | 1215 | |||
GenBank mRNA | AJ512753.3 | - | 6 | 984 | ||
AK090358.1 | - | 6 | 1181 | |||
AK145288.1 | - | 6 | 1216 | |||
AY189815.1 | - | 6 | 1123 | |||
AY189816.1 | - | 6 | 1123 | |||
BC121792.1 | - | 6 | 1054 | 40099393 |