Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177431.4 | NP_803180.3 | 35 | 5507 | ||
GenBank mRNA | AJ512753.3 | - | 35 | 5276 | ||
AY189815.1 | - | 35 | 5415 | |||
BC121792.1 | - | 35 | 5346 | 40099393 |