Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177611.3 | NP_808279.1 | 5 | 1411 | ||
GenBank mRNA | AK043669.1 | - | 6 | 1703 | ||
AK150238.1 | - | 5 | 1440 | |||
AK155249.1 | - | 5 | 1441 | |||
AK169859.1 | - | 5 | 1440 | |||
BC068231.1 | - | 5 | 1411 | 30361859 |