Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Os9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171026.1 | NP_001164497.1 | 12 | 1526 | ||
NM_177614.3 | NP_808282.2 | 12 | 1526 | |||
GenBank mRNA | AK028047.1 | - | 12 | 1502 | ||
AK179699.1 | - | 1 | 14 | |||
AK185854.1 | - | 1 | 14 | |||
BC006844.1 | - | 12 | 1346 | |||
BC031768.1 | - | 12 | 1491 | 4487829 | ||
CX566458.1 | - | 6 | 645 | |||
DV647242.1 | - | 2 | 370 |