Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177618.4 | NP_808286.2 | 3 | 874 | ||
GenBank mRNA | AK220346.1 | - | 2 | 580 | ||
BC030477.1 | - | 3 | 1012 | |||
BC048831.1 | - | 2 | 693 | 4504495 | ||
BC063070.1 | - | 3 | 874 | 5687975 |