Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161365.1 | NP_001154837.1 | 10 | 3476 | ||
NM_177620.4 | NP_808288.2 | 10 | 3587 | |||
GenBank mRNA | AK032205.1 | - | 10 | 3588 | ||
AK154506.1 | - | 10 | 3612 | |||
AK154522.1 | - | 10 | 3391 | |||
AK154963.1 | - | 10 | 3477 | |||
AK166196.1 | - | 10 | 3433 | |||
AK171301.1 | - | 10 | 3401 | |||
AK171311.1 | - | 10 | 3392 | |||
AY265356.1 | - | 10 | 3573 | |||
BC058432.1 | - | 45 | ||||
BC137907.1 | - | 10 | 3515 | 8860926 |