Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177632.3 | NP_808300.1 | 1 | 2301 | ||
GenBank mRNA | AK083036.1 | - | 1 | 2301 | ||
AK137950.1 | - | 1 | 2302 | |||
BC022623.1 | - | 1 | 894 | |||
BC026202.1 | - | 2 | 511 | |||
BC099947.1 | - | 1 | 2314 | 30088416 | ||
BC138882.1 | - | 1 | 1915 | 8861904 | ||
BC138884.1 | - | 1 | 1891 | 8861906 |