Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177664.5 | NP_808332.3 | 7 | 4499 | ||
GenBank mRNA | AF541279.1 | - | 7 | 4503 | ||
AK079360.1 | - | 1 | 1255 | |||
AK129149.1 | - | 7 | 4104 | |||
AY266266.1 | - | 7 | 4503 | |||
AY266267.1 | - | 7 | 4503 |