Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178045.4 | NP_835146.3 | 3, 4 | 276 | ||
GenBank mRNA | AK036528.1 | - | 3, 4 | 240 | ||
AK053578.1 | - | 3, 4 | 238 | |||
AK089460.1 | - | 3, 4 | 233 | |||
AK139284.1 | - | 3, 4 | 223 | |||
AK151561.1 | - | 3, 4 | 240 | |||
AK152639.1 | - | 3, 4 | 240 | |||
AK160584.1 | - | 3, 4 | 250 | |||
AK164357.1 | - | 3, 4 | 239 | |||
AK170955.1 | - | 3, 4 | 225 | |||
AK171198.1 | - | 3, 4 | 254 | |||
BC023245.1 | - | 3, 4 | 229 | 4459337 | ||
BC060709.1 | - | 3, 4 | 231 | 6409692 | ||
BY170346.1 | - | 3, 4 | 277 |