Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 5 | 577 | ||
NM_001190378.1 | NP_001177307.1 | 5 | 577 | |||
NM_001190379.1 | NP_001177308.1 | 5 | 577 | |||
NM_178116.4 | NP_835217.1 | 5 | 577 | |||
GenBank mRNA | AK039670.1 | - | 5 | 404 | ||
AK122400.1 | - | 3 | 233 | |||
AK150186.1 | - | 5 | 429 | |||
AK169118.1 | - | 5 | 416 | |||
AK195230.1 | - | 1 | 41 | |||
BC049133.1 | - | 5 | 406 | 6827800 | ||
BC056395.1 | - | 5 | 406 | 6852349 | ||
BC059065.1 | - | 4 | 305 |