Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289670.1 | NP_001276599.1 | 5 | 433 | ||
NM_001289682.1 | NP_001276611.1 | 6 | 516 | |||
NM_178252.3 | NP_839983.1 | 6 | 543 | |||
GenBank mRNA | AK142180.1 | - | 5 | 434 | ||
AY217764.1 | - | 5 | 376 | |||
BC065086.1 | - | 6 | 492 | 6850475 | ||
BC065166.1 | - | 6 | 490 | 6836510 | ||
BC066047.1 | - | 6 | 468 | 6822191 |