Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178367.4 | NP_848144.3 | 12 | 4063 | ||
GenBank mRNA | AK014230.1 | - | 1 | 809 | ||
BC027842.1 | - | 12 | 4059 | |||
BC052172.1 | - | 12 | 4056 | 6515901 |