Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN3 SIRNA Assays ›
Gene Symbol:
RTN3
Gene Name:
reticulon 3
Gene Aliases:
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1
Chromosome Location:
Chr.11: 63681315 - 63759891 on Build GRCh38
UniGene ID:
Hs.743229
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17160
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001265589.1
NP_001252518.1
4
2876
NM_001265590.1
NP_001252519.1
3
2540
NM_001265591.1
NP_001252520.1
2
488
NM_006054.3
NP_006045.1
2
488
NM_201428.2
NP_958831.1
3
2819
NM_201429.2
NP_958832.1
3
545
NM_201430.2
NP_958833.1
2
488
NR_049750.1
2
488
NR_049751.1
2
488
XM_011544730.2
XP_011543032.1
4
2966
XM_011544731.2
XP_011543033.1
3
680
GenBank mRNA
AB209771.1
-
2
425
AF059524.1
-
2
425
AF119297.1
-
2
437
AK075412.1
-
2
438
AK094965.1
-
2
1541
AK122727.1
-
2
941
AK127079.1
-
3
2515
AK222898.1
-
2
425
AK295361.1
-
3
450
AK295534.1
-
2
342
AK297529.1
-
2
365
AK297533.1
-
2
438
AK304100.1
-
4
2826
AK307811.1
-
3
2765
AK308083.1
-
2
433
AK309766.1
-
3
2494
AY427821.2
-
3
2786
AY750848.1
-
4
2843
BC000634.1
-
2
396
BC010556.1
-
2
401
BC011394.1
-
2
437
BC022993.1
-
2
427
BC100822.2
-
2
418
BC100823.2
-
2
418
BC105981.1
-
2
369
BC105982.1
-
2
400
BC118550.1
-
2
418
BC118628.1
-
2
418
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
chemical synaptic transmission
viral process
vesicle-mediated transport
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline