Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178676.4 | NP_848791.2 | 11 | 2135 | ||
GenBank mRNA | AK032057.1 | - | 11 | 2121 | ||
AK046218.1 | - | 11 | 2111 | |||
AK169947.1 | - | 6 | 1545 | |||
AK170974.1 | - | 6 | 1544 | |||
BC079871.1 | - | 11 | 2106 | 6831925 |