Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242419.1 | NP_001229348.1 | 7 | 2065 | ||
NM_178682.4 | NP_848797.1 | 6 | 2002 | |||
GenBank mRNA | AK030233.1 | - | 7 | 2088 | ||
AK054203.1 | - | 5 | 1717 | |||
AK129100.1 | - | 7 | 1828 | |||
AK143867.1 | - | 7 | 2051 | |||
AK150855.1 | - | 1 | 75 | |||
AK151439.1 | - | 1 | 75 | |||
AK151699.1 | - | 1 | 163 | |||
AK151939.1 | - | 6 | 1969 | |||
AK152357.1 | - | 1 | 75 | |||
AK152500.1 | - | 1 | 163 | |||
AK152820.1 | - | 6 | 1969 | |||
AK161132.1 | - | 6 | 2004 | |||
AK164823.1 | - | 6 | 1969 | |||
BC040401.1 | - | 6 | 1975 | 4193723 |