Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178690.4 | NP_848805.2 | 4 | 259 | ||
GenBank mRNA | AK076399.1 | - | 4 | 242 | ||
AK080432.1 | - | 4 | 241 | |||
AK134604.1 | - | 4 | 241 | |||
AK164523.1 | - | 4 | 240 | |||
AK170768.1 | - | 4 | 231 | |||
AK172881.1 | - | 4 | 246 | |||
BC046297.1 | - | 4 | 225 | 6486170 |