Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C130074G19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178692.3 | NP_848807.1 | 2 | 582 | ||
GenBank mRNA | AK052654.1 | - | 2 | 546 | ||
AK081755.1 | - | 2 | 561 | |||
AK084264.1 | - | 2 | 555 | |||
AK087640.1 | - | 2 | 573 | |||
BC116951.1 | - | 2 | 582 | 40126270 | ||
BC116953.1 | - | 2 | 582 | 40126272 |