Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145876.2 | NP_001139348.1 | 4 | 1396 | ||
NM_001145877.2 | NP_001139349.1 | 3 | 1314 | |||
NM_001281795.1 | NP_001268724.1 | 4 | 1317 | |||
NM_178696.5 | NP_848811.2 | 4 | 1442 | |||
GenBank mRNA | AK032960.1 | - | 4 | 1420 | ||
AK044648.1 | - | 5 | 1247 | |||
AK046586.1 | - | 4 | 1405 | |||
AK079532.1 | - | 4 | 1404 | |||
AK083273.1 | - | 4 | 1412 | |||
AK083470.1 | - | 4 | 1292 | |||
AK134914.1 | - | 4 | 1441 | |||
AK144731.1 | - | 4 | 1369 | |||
AK165159.1 | - | 3 | 1283 | |||
AK166011.1 | - | 4 | 1414 | |||
AK166331.1 | - | 4 | 1403 | |||
AK168955.1 | - | 4 | 1415 | |||
AK169886.1 | - | 4 | 1406 | |||
BC012234.1 | - | 4 | 1237 | 4191504 | ||
BC052771.1 | - | 4 | 1387 | 6310271 |