Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178697.4 | NP_848812.1 | 2 | 366 | ||
GenBank mRNA | AK028704.1 | - | 2 | 367 | ||
AK035512.1 | - | 2 | 433 | |||
AK048276.1 | - | 2 | 357 | |||
AY161007.1 | - | 2 | 322 | |||
BC096379.1 | - | 2 | 429 | 5317084 |