Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098476.2 | NP_001091946.1 | 5 | 614 | ||
NM_178700.5 | NP_848815.2 | 5 | 942 | |||
GenBank mRNA | AK044889.1 | - | 5 | 943 | ||
AK045986.1 | - | 5 | 896 | |||
AK077782.1 | - | 5 | 943 | |||
AK089014.1 | - | 4 | 478 | |||
AK168385.1 | - | 5 | 585 | |||
AK168459.1 | - | 5 | 585 | |||
AK210897.1 | - | 2 | 129 | |||
AK218546.1 | - | 2 | 129 |