Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098476.2 | NP_001091946.1 | 10 | 1852 | ||
NM_178700.5 | NP_848815.2 | 10 | 2180 | |||
GenBank mRNA | AK045986.1 | - | 10 | 2132 | ||
AK077782.1 | - | 10 | 2181 | |||
AK089014.1 | - | 9 | 1716 | |||
AK168385.1 | - | 10 | 1824 | |||
AK168459.1 | - | 10 | 1824 | |||
AK208573.1 | - | 1 | 14 | |||
AK211991.1 | - | 1 | 135 |