Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 1464 | ||
NM_178701.3 | NP_848816.3 | 3 | 1454 | |||
GenBank mRNA | AK046249.1 | - | 3 | 1463 | ||
AK050400.1 | - | 3 | 1424 | |||
AK080722.1 | - | 3 | 1398 | |||
AK137459.1 | - | 3 | 1465 | |||
AK167985.1 | - | 2 | 1172 | |||
AK206223.1 | - | 1 | 18 | |||
BC014756.1 | - | 1 | 488 | |||
BC037717.1 | - | 3 | 1435 | 4239389 |