Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202413.1 | NP_001189342.1 | 3, 4 | 382 | ||
NM_001202414.1 | NP_001189343.1 | 4, 5 | 510 | |||
NM_006066.3 | NP_006057.1 | 3, 4 | 599 | |||
NM_153326.2 | NP_697021.1 | 2, 3 | 471 | |||
GenBank mRNA | AK293083.1 | - | 2, 3 | 207 | ||
AK307068.1 | - | 2, 3 | 404 | |||
AK307237.1 | - | 2, 3 | 394 | |||
AU142853.1 | - | 3, 4 | 321 | |||
BC000670.2 | - | 3, 4 | 528 | 3349300 | ||
BC005394.1 | - | 2, 3 | 187 | 4051944 | ||
BI549149.1 | - | 2, 3 | 476 | |||
BT007003.1 | - | 1, 2 | 92 | |||
CR457010.1 | - | 1, 2 | 92 | |||
CX788871.1 | - | 4, 5 | 437 | |||
DA550374.1 | - | 2, 3 | 413 | |||
EU668319.1 | - | 2, 3 | 428 | |||
EU794588.1 | - | 2, 3 | 428 | |||
J04794.1 | - | 2, 3 | 152 |