Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178703.4 | NP_848818.1 | 8 | 1207 | ||
GenBank mRNA | AK042956.1 | - | 8 | 1069 | ||
AK047663.1 | - | 8 | 1053 | |||
AK052971.1 | - | 8 | 1032 | |||
AK053883.1 | - | 8 | 1029 | |||
BC059080.1 | - | 8 | 1058 | 6827121 | ||
M92378.1 | - | 7 | 966 |