Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178703.4 | NP_848818.1 | 16 | 3365 | ||
GenBank mRNA | AK042956.1 | - | 16 | 3227 | ||
AK053883.1 | - | 16 | 3187 | |||
BC027111.1 | - | 3 | 1561 | |||
BC059080.1 | - | 16 | 3216 | 6827121 |