Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178704.3 | NP_848819.2 | 5 | 638 | ||
GenBank mRNA | AF538322.1 | - | 5 | 622 | ||
AK031345.1 | - | 5 | 630 | |||
AK033395.1 | - | 5 | 595 | |||
AK034217.1 | - | 5 | 639 | |||
AK045758.1 | - | 5 | 539 | |||
BC050065.1 | - | 5 | 582 | 30016211 | ||
BC059897.1 | - | 5 | 535 | 6821433 |