Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 4, 5 | 1043 | ||
GenBank mRNA | AK032204.1 | - | 4, 5 | 700 | ||
AK034077.1 | - | 4, 5 | 661 | |||
AK034240.1 | - | 4, 5 | 695 | |||
AK082126.1 | - | 4, 5 | 791 | |||
AY164459.1 | - | 4, 5 | 1040 | |||
BC075646.1 | - | 4, 5 | 673 | 30620475 |